Assay Details
Target Gene Details
Entrez Gene ID: | 55471 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 7 |
Gene Aliases: |
C2orf56, MidA, PRO1853 |
Location: |
Chr.2:37231631-37259052 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFAF7 | NM_001083946.1 | NP_001077415.1 | ||
NM_144736.4 | NP_653337.1 | |||
XM_005264410.1 | XP_005264467.1 | |||
XM_005264411.1 | XP_005264468.1 | |||
XM_011532956.1 | XP_011531258.1 | |||
AK294240.1 | ||||
AK299015.1 | ||||
AK302150.1 | ||||
AK303555.1 | ||||
BC004548.2 | AAH04548.2 | |||
BI334629.1 | ||||
BM172474.1 | ||||
BX538031.1 | CAD97976.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2684 | Chr.2:37203356 - 37236290 on Build GRCh38 | Insertion | CEBPZ NDUFAF7 CEBPZOS |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |