Assay Details
Target Gene Details
Entrez Gene ID: | 101927492 |
Gene Name: | uncharacterized LOC101927492 |
Gene Aliases: |
- |
Location: |
Chr.2:105097052-105102944 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UTAT33 | NR_110227.1 | |||
CR736606.1 |