Assay Details
Target Gene Details
Entrez Gene ID: | 27178 |
Gene Name: | interleukin 37 |
Gene Aliases: |
FIL1, FIL1(ZETA), FIL1Z, IL-1F7, IL-1H, IL-1H4, IL-1RP1, IL-37, IL1F7, IL1H4, IL1RP1 |
Location: |
Chr.2:112908889-112918882 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IL37 | XM_011510963.2 | XP_011509265.1 | ||
XM_011510964.2 | XP_011509266.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3584019 | Chr.2:112910229 - 112918462 on Build GRCh38 | Loss |
![]() |
nsv2879 | Chr.2:112882915 - 112916265 on Build GRCh38 | Insertion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)