Assay Details
Target Gene Details
Entrez Gene ID: | 54884 |
Gene Name: | retinol saturase |
Gene Aliases: |
- |
Location: |
Chr.2:85341955-85354698 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RETSAT | NM_017750.3 | NP_060220.3 | ||
AK075261.1 | BAC11505.1 | |||
AK296522.1 | ||||
AL833237.1 | ||||
AY358568.1 | AAQ88931.1 | |||
BC068517.1 | AAH68517.1 | |||
DA418052.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3575266 | Chr.2:85065061 - 85379120 on Build GRCh38 | Gain | LOC102724579 ELMOD3 TCF7L1 RETSAT LOC100996478 TGOLN2 |
nsv834282 | Chr.2:85237271 - 85414189 on Build GRCh38 | Gain | ELMOD3 CAPG TCF7L1 RETSAT TGOLN2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |