Assay Details
Target Gene Details
Entrez Gene ID: | 9392 |
Gene Name: | transforming growth factor beta receptor associated protein 1 |
Gene Aliases: |
TRAP-1, TRAP1, VPS3 |
Location: |
Chr.2:105249404-105329714 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TGFBRAP1 | XM_011512193.2 | XP_011510495.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1012462 | Chr.2:105183092 - 105294801 on Build GRCh38 | Gain | TGFBRAP1 GPR45 |
nsv582577 | Chr.2:105167153 - 105302140 on Build GRCh38 | Loss | TGFBRAP1 GPR45 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |