Assay Details
Target Gene Details
Entrez Gene ID: | 3176 |
Gene Name: | histamine N-methyltransferase |
Gene Aliases: |
HMT, HNMT-S1, HNMT-S2, MRT51 |
Location: |
Chr.2:137964068-138016364 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HNMT | NM_006895.2 | NP_008826.1 | ||
XM_011511064.2 | XP_011509366.1 | |||
XM_017003948.1 | XP_016859437.1 | |||
XM_017003949.1 | XP_016859438.1 | |||
AF523358.1 | AAN33016.1 | |||
AF523359.1 | AAN33017.1 | |||
AF523360.1 | AAN33018.1 | |||
AK301462.1 | ||||
AK313804.1 | ||||
BC020677.1 | AAH20677.1 | |||
D16224.1 | BAA03752.1 | |||
U08092.1 | AAA17423.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv583180 | Chr.2:137597152 - 138007481 on Build GRCh38 | Gain | THSD7B LOC101928273 HNMT |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |