Assay Details
Target Gene Details
Entrez Gene ID: | 79623 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 14 |
Gene Aliases: |
GALNT15, GalNac-T10, GalNac-T14 |
Location: |
Chr.2:30888518-31138726 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GALNT14 | NM_001253826.1 | NP_001240755.1 | ||
NM_001253827.1 | NP_001240756.1 | |||
NM_024572.3 | NP_078848.2 | |||
XM_011533105.1 | XP_011531407.1 | |||
XM_017004906.1 | XP_016860395.1 | |||
XM_017004907.1 | XP_016860396.1 | |||
XM_017004908.1 | XP_016860397.1 | |||
AB078144.1 | BAC56889.1 | |||
AJ505966.1 | CAD44543.1 | |||
AK022753.1 | BAB14227.1 | |||
AK074394.1 | ||||
AK091313.1 | BAC03634.1 | |||
AK122747.1 | ||||
AK225808.1 | ||||
AK298735.1 | ||||
AY358758.1 | AAQ89118.1 | |||
BC006269.2 | AAH06269.2 | |||
BC010659.1 | AAH10659.1 | |||
Y09324.4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833659 | Chr.2:30797706 - 30948639 on Build GRCh38 | Gain | GALNT14 CAPN13 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |