Assay Details
Target Gene Details
Entrez Gene ID: | 390 |
Gene Name: | Rho family GTPase 3 |
Gene Aliases: |
ARHE, Rho8, RhoE, memB |
Location: |
Chr.2:150468193-150487695 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RND3 | NM_001254738.1 | NP_001241667.1 | ||
NM_005168.4 | NP_005159.1 | |||
AF498969.1 | AAM21116.1 | |||
AK298007.1 | ||||
AK299731.1 | ||||
AK309442.1 | ||||
AK311312.1 | ||||
AK313224.1 | ||||
AL702945.1 | ||||
BC012513.1 | AAH12513.1 | |||
BQ440721.1 | ||||
BT006769.1 | AAP35415.1 | |||
CR542038.1 | CAG46835.1 | |||
CR542070.1 | CAG46867.1 | |||
DC367501.1 | ||||
S82240.1 | AAB47133.1 | |||
X95282.1 | CAA64603.1 | |||
X97758.1 | CAA66352.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834414 | Chr.2:150352860 - 150556795 on Build GRCh38 | Gain | RND3 LOC101929260 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |