Assay Details
Target Gene Details
Entrez Gene ID: | 374987 |
Gene Name: | NEXN antisense RNA 1 |
Gene Aliases: |
C1orf118 |
Location: |
Chr.1:77881348-77889539 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NEXN-AS1 | NR_103535.1 | |||
AK075118.1 | BAC11413.1 |