Assay Details
Target Gene Details
Entrez Gene ID: | 80342 |
Gene Name: | TRAF3 interacting protein 3 |
Gene Aliases: |
T3JAM |
Location: |
Chr.1:209756032-209782323 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TRAF3IP3 | NM_001320144.1 | NP_001307073.1 | ||
NM_025228.3 | NP_079504.2 | |||
NR_109871.1 | ||||
XM_011510018.2 | XP_011508320.1 | |||
XM_011510019.1 | XP_011508321.1 | |||
XM_017002399.1 | XP_016857888.1 | |||
XM_017002400.1 | XP_016857889.1 | |||
AK122792.1 | ||||
AK225762.1 | ||||
AK300555.1 | ||||
AK312755.1 | ||||
AL049667.1 | CAB41242.1 | |||
AL558384.3 | ||||
BC110302.1 | AAI10303.1 | |||
BX648450.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv4343 | Chr.1:209756587 - 209801532 on Build GRCh38 | Deletion | IRF6 TRAF3IP3 C1orf74 |
nsv1006632 | Chr.1:209712857 - 209797010 on Build GRCh38 | Gain | IRF6 LOC101930114 TRAF3IP3 HSD11B1 C1orf74 |
nsv1001588 | Chr.1:209735531 - 209764898 on Build GRCh38 | Gain | LOC101930114 TRAF3IP3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |