Assay Details
Target Gene Details
Entrez Gene ID: | 6512 |
Gene Name: | solute carrier family 1 member 7 |
Gene Aliases: |
AAAT, EAAT5 |
Location: |
Chr.1:53087179-53142632 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC1A7 | NM_001287595.1 | NP_001274524.1 | ||
NM_001287597.1 | NP_001274526.1 | |||
NM_006671.5 | NP_006662.3 | |||
NR_109858.1 | ||||
AB593059.1 | ||||
AB593060.1 | ||||
AB593061.1 | ||||
BC012119.1 | AAH12119.1 | |||
BC017242.2 | AAH17242.1 | |||
JF917096.1 | ||||
U76362.1 | AAB53971.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1003159 | Chr.1:53079580 - 53117512 on Build GRCh38 | Gain |
![]() ![]() |
nsv1010121 | Chr.1:52939995 - 53134678 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv546240 | Chr.1:53084515 - 53134406 on Build GRCh38 | Loss |
![]() ![]() |
nsv950739 | Chr.1:53061429 - 53136228 on Build GRCh38 | Deletion |
![]() ![]() |
nsv522862 | Chr.1:53038739 - 53102049 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv516780 | Chr.1:53054454 - 53102049 on Build GRCh38 | Gain+Loss |
![]() ![]() |
nsv1001535 | Chr.1:52620368 - 53346539 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv88e59 | Chr.1:53094492 - 53102090 on Build GRCh38 | Duplication |
![]() |
nsv1115343 | Chr.1:53097228 - 53102228 on Build GRCh38 | Duplication |
![]() |
esv21434 | Chr.1:53094968 - 53103242 on Build GRCh38 | Gain+Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)