Assay Details
Target Gene Details
Entrez Gene ID: | 64326 |
Gene Name: | ring finger and WD repeat domain 2 |
Gene Aliases: |
COP1, RNF200 |
Location: |
Chr.1:175944826-176207279 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RFWD2 | NM_001001740.3 | NP_001001740.1 | ||
NM_001286644.1 | NP_001273573.1 | |||
NM_022457.6 | NP_071902.2 | |||
XM_005245447.2 | XP_005245504.1 | |||
XM_005245448.2 | XP_005245505.1 | |||
XM_006711487.2 | XP_006711550.1 | |||
XM_011509881.1 | XP_011508183.1 | |||
XM_017002076.1 | XP_016857565.1 | |||
AF508940.1 | AAM34692.1 | |||
AF527539.1 | AAQ08989.1 | |||
AK001278.1 | ||||
AK314750.1 | ||||
AY509921.1 | AAS82851.1 | |||
BC094728.1 | AAH94728.1 | |||
BX537932.1 | ||||
CA450176.1 | ||||
KJ534928.1 | ||||
KJ534929.1 | ||||
KJ534930.1 | ||||
KJ534931.1 | ||||
KJ534932.1 | ||||
KJ534933.1 | ||||
KJ534992.1 | ||||
KJ535061.1 | ||||
KJ535076.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)