Assay Details
Target Gene Details
Entrez Gene ID: | 5928 |
Gene Name: | RB binding protein 4, chromatin remodeling factor |
Gene Aliases: |
NURF55, RBAP48, lin-53 |
Location: |
Chr.1:32651148-32686211 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RBBP4 | NM_001135255.1 | NP_001128727.1 | ||
NM_001135256.1 | NP_001128728.1 | |||
NM_005610.2 | NP_005601.1 | |||
AK222779.1 | BAD96499.1 | |||
AK299251.1 | ||||
AK299410.1 | ||||
AK312571.1 | ||||
BC003092.1 | AAH03092.1 | |||
BC015123.1 | AAH15123.1 | |||
BC053904.1 | AAH53904.1 | |||
BC075836.1 | AAH75836.1 | |||
BT007309.1 | AAP35973.1 | |||
X71810.1 | CAA50685.1 | |||
X74262.1 | CAA52321.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv545961 | Chr.1:32566811 - 32685350 on Build GRCh38 | Loss | ZBTB8A RBBP4 SYNC ZBTB8OS |
nsv519188 | Chr.1:32619643 - 32780839 on Build GRCh38 | Loss | KIAA1522 RBBP4 YARS SYNC ZBTB8OS |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |