Assay Details
Target Gene Details
Entrez Gene ID: | 3918 |
Gene Name: | laminin subunit gamma 2 |
Gene Aliases: |
B2T, BM600, CSF, EBR2, EBR2A, LAMB2T, LAMNB2 |
Location: |
Chr.1:183186039-183245127 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LAMC2 | NM_005562.2 | NP_005553.2 | ||
NM_018891.2 | NP_061486.2 | |||
XM_017001273.1 | XP_016856762.1 | |||
AK296944.1 | ||||
AK307945.1 | ||||
BC112286.1 | AAI12287.1 | |||
BC113378.1 | ||||
X73902.1 | CAA52108.1 | |||
Z15008.1 | CAA78728.1 | |||
Z15009.1 | CAA78729.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv548375 | Chr.1:183151893 - 183208319 on Build GRCh38 | Gain | LAMC2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |