Assay Details
Target Gene Details
Entrez Gene ID: | 51097 |
Gene Name: | saccharopine dehydrogenase (putative) |
Gene Aliases: |
CGI-49, NET11 |
Location: |
Chr.1:246724076-246768137 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SCCPDH | NM_016002.2 | NP_057086.2 | ||
AF151807.1 | AAD34044.1 | |||
AK075178.1 | BAC11453.1 | |||
BC026185.2 | AAH26185.1 | |||
BG258610.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv523531 | Chr.1:246730646 - 246804597 on Build GRCh38 | Gain | SCCPDH LINC01341 |
esv2667587 | Chr.1:246652751 - 246920518 on Build GRCh38 | Deletion | AHCTF1 SCCPDH LOC102724382 CNST LINC01341 |
nsv1006262 | Chr.1:246729524 - 246801473 on Build GRCh38 | Gain | SCCPDH LINC01341 |
esv2762192 | Chr.1:246660555 - 246905930 on Build GRCh38 | Gain | AHCTF1 SCCPDH LOC102724382 CNST LINC01341 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |