Assay Details
Target Gene Details
Entrez Gene ID: | 4678 |
Gene Name: | nuclear autoantigenic sperm protein |
Gene Aliases: |
FLB7527, HMDRA1, PRO1999 |
Location: |
Chr.1:45583988-45618906 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NASP | NM_001195193.1 | NP_001182122.1 | ||
NM_002482.3 | NP_002473.2 | |||
NM_152298.3 | NP_689511.2 | |||
XM_005270888.2 | XP_005270945.1 | |||
XM_005270889.2 | XP_005270946.1 | |||
XM_011541509.1 | XP_011539811.1 | |||
XM_017001356.1 | XP_016856845.1 | |||
AF035191.1 | AAD02006.1 | |||
AK056161.1 | ||||
AK222778.1 | BAD96498.1 | |||
AK222816.1 | BAD96536.1 | |||
AK291637.1 | ||||
AK298893.1 | ||||
AK299584.1 | ||||
AK308001.1 | ||||
BC003113.1 | AAH03113.1 | |||
BC009933.2 | AAH09933.1 | |||
BC010105.2 | AAH10105.1 | |||
BC011580.1 | AAH11580.1 | |||
BC053608.1 | AAH53608.1 | |||
BT006757.1 | AAP35403.1 | |||
DC361634.1 | ||||
M97856.1 | AAA36027.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893512 | Chr.1:45476180 - 46068871 on Build GRCh38 | Gain | NASP AKR1A1 PIK3R3 PRDX1 RPS15AP10 GPBP1L1 CCDC17 IPP MAST2 TESK2 TMEM69 CCDC163P MMACHC |
nsv829726 | Chr.1:45431909 - 45637514 on Build GRCh38 | Loss | CCDC17 TESK2 NASP AKR1A1 PRDX1 GPBP1L1 CCDC163P MMACHC |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |