Assay Details
Target Gene Details
Entrez Gene ID: | 11124 |
Gene Name: | Fas associated factor 1 |
Gene Aliases: |
CGI-03, HFAF1s, UBXD12, UBXN3A, hFAF1 |
Location: |
Chr.1:50441263-50960264 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAF1 | NM_007051.2 | NP_008982.1 | ||
XM_017000136.1 | XP_016855625.1 | |||
AF094700.1 | AAD51876.1 | |||
AF106798.1 | AAD51886.1 | |||
AF132938.1 | AAD27713.1 | |||
AF136173.1 | AAP97263.1 | |||
AJ271408.1 | CAB67705.1 | |||
AK094843.1 | ||||
AK292840.1 | ||||
AK293659.1 | ||||
AL133631.1 | CAB63755.1 | |||
BC004970.1 | AAH04970.1 | |||
BC067100.1 | AAH67100.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)