Assay Details
Target Gene Details
Entrez Gene ID: | 257194 |
Gene Name: | neuronal growth regulator 1 |
Gene Aliases: |
DMML2433, IGLON4, KILON, Ntra |
Location: |
Chr.1:71402942-72282850 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NEGR1 | NM_173808.2 | NP_776169.2 | ||
AK092307.1 | BAC03858.1 | |||
AK295481.1 | ||||
BC036771.1 | AAH36771.1 | |||
BX538014.1 | CAD97961.1 | |||
BX648027.1 | ||||
CR749215.1 | CAH18072.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830170 | Chr.1:71390764 - 71543075 on Build GRCh38 | Gain | NEGR1 |
nsv830159 | Chr.1:71306625 - 71444177 on Build GRCh38 | Gain | NEGR1 |
nsv546475 | Chr.1:71383354 - 71472011 on Build GRCh38 | Gain | NEGR1 |
nsv546474 | Chr.1:71327538 - 71574671 on Build GRCh38 | Gain | NEGR1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |