Assay Details
Target Gene Details
Entrez Gene ID: | 81493 |
Gene Name: | syncoilin, intermediate filament protein |
Gene Aliases: |
SYNC1, SYNCOILIN |
Location: |
Chr.1:32679906-32702760 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SYNC | NM_001161708.1 | NP_001155180.1 | ||
NM_030786.2 | NP_110413.2 | |||
AK297958.1 | ||||
AK302275.1 | ||||
DC355889.1 | ||||
DC398945.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509158 | Chr.1:32691713 - 32783820 on Build GRCh38 | Insertion | SYNC KIAA1522 YARS |
nsv519188 | Chr.1:32619643 - 32780839 on Build GRCh38 | Loss | ZBTB8OS SYNC KIAA1522 RBBP4 YARS |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |