Assay Details
Target Gene Details
Entrez Gene ID: | 10446 |
Gene Name: | leucine rich repeat neuronal 2 |
Gene Aliases: |
FIGLER7, GAC1, LRANK1, LRRN5 |
Location: |
Chr.1:204617170-204685836 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRRN2 | NM_006338.2 | NP_006329.2 | ||
NM_201630.1 | NP_963924.1 | |||
XM_005244827.3 | XP_005244884.1 | |||
XM_011509066.2 | XP_011507368.1 | |||
XM_017000050.1 | XP_016855539.1 | |||
AF030435.1 | AAC39792.1 | |||
AK226055.1 | ||||
AK312408.1 | ||||
BC034047.1 | AAH34047.1 | |||
BC068541.1 | AAH68541.1 | |||
T08293.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3588622 | Chr.1:204585680 - 204625853 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)