Assay Details
Target Gene Details
Entrez Gene ID: | 10390 |
Gene Name: | choline/ethanolamine phosphotransferase 1 |
Gene Aliases: |
- |
Location: |
Chr.1:111139436-111185102 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CEPT1 | NM_001007794.1 | NP_001007795.1 | ||
NM_006090.3 | NP_006081.1 | |||
XM_006710285.3 | XP_006710348.1 | |||
XM_011540496.1 | XP_011538798.1 | |||
AF068302.1 | AAD25170.1 | |||
AF138862.1 | AAF61194.1 | |||
AK023381.1 | ||||
BC032610.2 | AAH32610.1 | |||
BC049196.1 | AAH49196.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv10639 | Chr.1:111126227 - 111244431 on Build GRCh38 | Gain | CHI3L2 DRAM2 CEPT1 DENND2D |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |