Assay Details
Target Gene Details
Entrez Gene ID: | 8564 |
Gene Name: | kynurenine 3-monooxygenase (kynurenine 3-hydroxylase) |
Gene Aliases: |
dJ317G22.1 |
Location: |
Chr.1:241531883-241595647 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KMO | NM_003679.4 | NP_003670.2 | ||
XM_005273337.1 | XP_005273394.1 | |||
XM_005273338.3 | XP_005273395.1 | |||
XM_017002649.1 | XP_016858138.1 | |||
AF056032.1 | AAC62615.1 | |||
AK291558.1 | ||||
AK310937.1 | ||||
AU137977.1 | ||||
BC005297.1 | AAH05297.1 | |||
BP276365.1 | ||||
Y13153.1 | CAA73613.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv549460 | Chr.1:241331278 - 241669985 on Build GRCh38 | Gain | FH CHML RGS7 KMO WDR64 OPN3 |
nsv1002214 | Chr.1:241295639 - 241936488 on Build GRCh38 | Gain | FH CHML RGS7 EXO1 KMO WDR64 OPN3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |