Assay Details
Target Gene Details
Entrez Gene ID: | 5906 |
Gene Name: | RAP1A, member of RAS oncogene family |
Gene Aliases: |
C21KG, G-22K, KREV-1, KREV1, RAP1, SMGP21 |
Location: |
Chr.1:111542223-111716695 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAP1A | XM_017001964.1 | XP_016857453.1 | ||
M22995.1 | AAA36150.1 |
Target Gene Details
Entrez Gene ID: | 57413 |
Gene Name: | transmembrane and immunoglobulin domain containing 3 |
Gene Aliases: |
AD026 |
Location: |
Chr.1:111483348-111564003 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMIGD3 | NM_001081976.2 | NP_001075445.1 | ||
NM_001302680.1 | NP_001289609.1 | |||
BC041707.1 | AAH41707.1 | |||
BC064411.1 | AAH64411.1 | |||
BI828255.1 | ||||
HY011888.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2421 | Chr.1:111521152 - 111567287 on Build GRCh38 | Deletion | RAP1A TMIGD3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |