Assay Details
Target Gene Details
Entrez Gene ID: | 23528 |
Gene Name: | zinc finger protein 281 |
Gene Aliases: |
ZBP-99, ZNP-99 |
Location: |
Chr.1:200404947-200483604 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF281 | XM_017000887.1 | XP_016856376.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3588558 | Chr.1:200423958 - 200429194 on Build GRCh38 | Loss |
![]() |
nsv548990 | Chr.1:200400630 - 200452338 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)