Assay Details
Target Gene Details
Entrez Gene ID: | 1912 |
Gene Name: | polyhomeotic homolog 2 |
Gene Aliases: |
EDR2, HPH2, PH2 |
Location: |
Chr.1:33323623-33431017 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834613 | Chr.1:33334605 - 33496188 on Build GRCh38 | Gain | ZSCAN20 LOC101929464 PHC2 |
nsv516693 | Chr.1:33319026 - 33354533 on Build GRCh38 | Loss | MIR3605 LOC101929464 PHC2 A3GALT2 |
nsv522364 | Chr.1:33266591 - 33367332 on Build GRCh38 | Loss | MIR3605 LOC101929464 PHC2 ZNF362 A3GALT2 |
nsv545963 | Chr.1:33254041 - 33354533 on Build GRCh38 | Loss | MIR3605 LOC101929464 PHC2 ZNF362 A3GALT2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |