Assay Details
Target Gene Details
Entrez Gene ID: | 2023 |
Gene Name: | enolase 1 |
Gene Aliases: |
ENO1L1, HEL-S-17, MPB1, NNE, PPH |
Location: |
Chr.1:8861000-8879092 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENO1 | NM_001201483.1 | NP_001188412.1 | ||
NM_001428.3 | NP_001419.1 | |||
XM_006710433.2 | XP_006710496.1 | |||
AF035286.1 | AAB88178.1 | |||
AK098420.1 | ||||
AK222517.1 | BAD96237.1 | |||
AK223192.1 | BAD96912.1 | |||
AK315417.1 | ||||
AL833741.1 | CAH56247.1 | |||
BC001810.2 | AAH01810.1 | |||
BC004325.1 | AAH04325.1 | |||
BC004458.1 | AAH04458.1 | |||
BC009218.2 | AAH09218.2 | |||
BC009912.2 | AAH09912.1 | |||
BC011130.1 | AAH11130.1 | |||
BC015641.2 | AAH15641.1 | |||
BC021166.2 | AAH21166.2 | |||
BC022545.1 | AAH22545.1 | |||
BC027725.2 | AAH27725.1 | |||
BC050642.2 | AAH50642.1 | |||
BC113034.1 | ||||
BT007163.1 | AAP35827.1 | |||
BX537400.1 | CAD97642.1 | |||
GQ472226.1 | ||||
GU170215.1 | ||||
M14328.1 | AAA52387.1 | |||
M55914.1 | AAA35698.1 | |||
X84907.1 | CAA59331.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv545345 | Chr.1:8831066 - 9035551 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv1001865 | Chr.1:8843872 - 8873183 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)