Assay Details
Target Gene Details
Entrez Gene ID: | 7203 |
Gene Name: | chaperonin containing TCP1 subunit 3 |
Gene Aliases: |
CCT-gamma, CCTG, PIG48, TCP-1-gamma, TRIC5 |
Location: |
Chr.1:156308961-156338415 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCT3 | NM_001008800.2 | NP_001008800.1 | ||
NM_005998.4 | NP_005989.3 | |||
NR_036564.1 | ||||
NR_036565.1 | ||||
AB208882.1 | BAD92119.1 | |||
AK126400.1 | ||||
AK293477.1 | ||||
AK300765.1 | ||||
AK308023.1 | ||||
AK315923.1 | ||||
AK316387.1 | ||||
AL518070.3 | ||||
AL833197.1 | CAI46192.1 | |||
AY524046.1 | AAS94255.1 | |||
BC006501.2 | ||||
BC008019.1 | AAH08019.1 | |||
BG476176.1 | ||||
BG746513.1 | ||||
X74801.1 | CAA52808.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)