Assay Details
Target Gene Details
Entrez Gene ID: | 54874 |
Gene Name: | formin binding protein 1 like |
Gene Aliases: |
C1orf39, TOCA1 |
Location: |
Chr.1:93448067-93569671 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FNBP1L | NM_001024948.2 | NP_001020119.1 | ||
NM_001164473.2 | NP_001157945.1 | |||
NM_017737.4 | NP_060207.2 | |||
XM_011541625.2 | XP_011539927.1 | |||
XM_017001533.1 | XP_016857022.1 | |||
AY514449.1 | AAR98814.1 | |||
DW423801.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)