Assay Details
Target Gene Details
Entrez Gene ID: | 59352 |
Gene Name: | leucine rich repeat containing G protein-coupled receptor 6 |
Gene Aliases: |
GPCR, VTS20631 |
Location: |
Chr.1:202193990-202319761 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LGR6 | NM_001017403.1 | NP_001017403.1 | ||
AB049405.1 | BAB39854.1 | |||
BM699180.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1010102 | Chr.1:202164648 - 202260980 on Build GRCh38 | Gain | LGR6 PTPRVP |
nsv522841 | Chr.1:202150606 - 202311878 on Build GRCh38 | Gain | LGR6 PTPRVP PTPN7 |
esv3891059 | Chr.1:202184455 - 202231671 on Build GRCh38 | Gain | LGR6 PTPRVP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |