Assay Details
Target Gene Details
Entrez Gene ID: | 57685 |
Gene Name: | cache domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.1:64470793-64693058 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CACHD1 | NM_020925.2 | NP_065976.2 | ||
CX163220.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1122355 | Chr.1:64477933 - 64492378 on Build GRCh38 | Deletion | CACHD1 |
nsv546437 | Chr.1:64230394 - 64538095 on Build GRCh38 | Loss | CACHD1 UBE2U |
esv2660804 | Chr.1:64381364 - 64626908 on Build GRCh38 | Deletion | CACHD1 MIR4794 |
esv3586265 | Chr.1:64420465 - 64489064 on Build GRCh38 | Loss | CACHD1 |
nsv830048 | Chr.1:64367523 - 64545238 on Build GRCh38 | Gain | CACHD1 |
nsv1000188 | Chr.1:64263100 - 64546875 on Build GRCh38 | Gain | CACHD1 UBE2U |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |