Assay Details
Target Gene Details
Entrez Gene ID: | 2326 |
Gene Name: | flavin containing monooxygenase 1 |
Gene Aliases: |
- |
Location: |
Chr.1:171248471-171285978 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FMO1 | NM_001282693.1 | NP_001269622.1 | ||
NM_001282694.1 | NP_001269623.1 | |||
NM_002021.2 | NP_002012.1 | |||
XM_005245038.3 | XP_005245095.1 | |||
XM_006711241.3 | XP_006711304.1 | |||
XM_006711242.3 | XP_006711305.1 | |||
AK290113.1 | ||||
AK296198.1 | ||||
AK315100.1 | ||||
DA537896.1 | ||||
M64082.1 | AAA52457.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831882 | Chr.1:171197085 - 171356944 on Build GRCh38 | Loss | FMO2 FMO1 FMO4 TOP1P1 |
esv28731 | Chr.1:171046945 - 171757821 on Build GRCh38 | Gain+Loss | VAMP4 FMO2 MIR1295A FMO1 FMO4 PRRC2C MIR1295B MYOC FMO3 FMO6P TOP1P1 MROH9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |