Assay Details
Target Gene Details
Entrez Gene ID: | 57413 |
Gene Name: | transmembrane and immunoglobulin domain containing 3 |
Gene Aliases: |
AD026 |
Location: |
Chr.1:111483348-111564003 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMIGD3 | NM_001081976.2 | NP_001075445.1 | ||
NM_001302680.1 | NP_001289609.1 | |||
NM_020683.6 | NP_065734.5 | |||
AY358644.1 | AAQ89007.1 | |||
BC041707.1 | AAH41707.1 | |||
BC064411.1 | AAH64411.1 | |||
BI828255.1 | ||||
HY011888.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv463250 | Chr.1:111195294 - 111519590 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv2410 | Chr.1:111486928 - 111510250 on Build GRCh38 | Insertion |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)