Assay Details
Target Gene Details
Entrez Gene ID: | 22866 |
Gene Name: | connector enhancer of kinase suppressor of Ras 2 |
Gene Aliases: |
CNK2, KSR2, MAGUIN |
Location: |
Chr.X:21374335-21654695 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CNKSR2 | NM_001168647.1 | NP_001162118.1 | ||
NM_001168648.1 | NP_001162119.1 | |||
NM_001168649.1 | NP_001162120.1 | |||
NM_014927.3 | NP_055742.2 | |||
XM_011545471.2 | XP_011543773.1 | |||
XM_011545472.2 | XP_011543774.1 | |||
XM_017029358.1 | XP_016884847.1 | |||
AB020709.1 | BAA74925.2 | |||
AF418269.1 | AAL60502.1 | |||
AF418270.1 | AAL60503.1 | |||
AK294728.1 | ||||
BC126121.1 | ||||
BC136289.1 | ||||
BC143839.1 | ||||
BP200041.1 | ||||
DA067208.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)