Assay Details
Target Gene Details
Entrez Gene ID: | 5160 |
Gene Name: | pyruvate dehydrogenase (lipoamide) alpha 1 |
Gene Aliases: |
PDHA, PDHAD, PDHCE1A, PHE1A |
Location: |
Chr.X:19343893-19361707 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDHA1 | NM_000284.3 | NP_000275.1 | ||
NM_001173454.1 | NP_001166925.1 | |||
NM_001173455.1 | NP_001166926.1 | |||
NM_001173456.1 | NP_001166927.1 | |||
XM_017029574.1 | XP_016885063.1 | |||
AK129887.1 | ||||
AK222740.1 | BAD96460.1 | |||
AK222988.1 | BAD96708.1 | |||
AK293250.1 | ||||
AK296341.1 | ||||
AK296457.1 | ||||
AK312263.1 | ||||
BC002406.1 | AAH02406.1 | |||
DC361191.1 | ||||
J03503.1 | AAA60055.1 | |||
J03575.1 | AAA60050.1 | |||
L13318.1 | AAA60227.1 | |||
L48690.1 | AAB59581.1 | |||
M24848.1 | AAA36533.1 | |||
X52709.1 | CAA36933.1 | |||
X52710.1 | CAA36934.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73453457] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |