Assay Details
Target Gene Details
Entrez Gene ID: | 51360 |
Gene Name: | membrane bound transcription factor peptidase, site 2 |
Gene Aliases: |
BRESEK, IFAP, KFSD, KFSDX, OLMSX, S2P |
Location: |
Chr.X:21839538-21885423 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MBTPS2 | NM_015884.3 | NP_056968.1 | ||
AF019612.1 | AAC51937.1 | |||
AK292933.1 | ||||
AK307143.1 | ||||
AK312474.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |