Assay Details
Target Gene Details
Entrez Gene ID: | 4810 |
Gene Name: | NHS actin remodeling regulator |
Gene Aliases: |
CTRCT40, CXN, SCML1 |
Location: |
Chr.X:17375420-17735994 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NHS | NM_001291867.1 | NP_001278796.1 | ||
NM_198270.3 | NP_938011.1 | |||
AK302437.1 | ||||
AY436752.1 | AAR03104.1 | |||
BC136415.1 | ||||
BC171763.1 | ||||
GQ988776.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv503n21 | Chr.X:17329494 - 17542854 on Build GRCh38 | Gain | NHS LOC101928389 MIR4768 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |