Assay Details
Target Gene Details
Entrez Gene ID: | 340554 |
Gene Name: | zinc finger CCCH-type containing 12B |
Gene Aliases: |
CXorf32, MCPIP2 |
Location: |
Chr.X:65034815-65507887 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZC3H12B | XM_017029479.1 | XP_016884968.1 | ||
XM_017029484.1 | XP_016884973.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |