Assay Details
Target Gene Details
Entrez Gene ID: | 1831 |
Gene Name: | TSC22 domain family member 3 |
Gene Aliases: |
DIP, DSIPI, GILZ, TSC-22R |
Location: |
Chr.X:107713221-107777329 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSC22D3 | NM_001318468.1 | NP_001305397.1 | ||
NM_001318470.1 | NP_001305399.1 | |||
NM_198057.2 | NP_932174.1 | |||
XM_005262099.1 | XP_005262156.1 | |||
XM_005262100.1 | XP_005262157.1 | |||
XM_005262102.1 | XP_005262159.1 | |||
XM_005262103.3 | XP_005262160.1 | |||
XM_011530884.1 | XP_011529186.1 | |||
XM_017029335.1 | XP_016884824.1 | |||
AK092645.1 | BAC03934.1 | |||
AK092669.1 | ||||
AK300535.1 | ||||
BM047061.1 | ||||
BX647854.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |