Assay Details
Target Gene Details
Entrez Gene ID: | 139378 |
Gene Name: | adhesion G protein-coupled receptor G4 |
Gene Aliases: |
GPR112, PGR17, RP1-299I16 |
Location: |
Chr.X:136300963-136416888 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADGRG4 | NM_153834.3 | NP_722576.3 | ||
XM_011531269.2 | XP_011529571.1 | |||
XM_011531271.2 | XP_011529573.1 | |||
AY882585.1 | AAW78659.1 | |||
CR936801.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |