Assay Details
Target Gene Details
Entrez Gene ID: | 27035 |
Gene Name: | NADPH oxidase 1 |
Gene Aliases: |
GP91-2, MOX1, NOH-1, NOH1 |
Location: |
Chr.X:100843324-100874345 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NOX1 | NM_001271815.1 | NP_001258744.1 | ||
NM_007052.4 | NP_008983.2 | |||
NM_013955.2 | NP_039249.1 | |||
AF166326.1 | AAF23232.1 | |||
AF166327.1 | AAF23233.1 | |||
AF166328.1 | AAF23234.1 | |||
AJ438989.1 | CAD27735.1 | |||
AK292201.1 | ||||
AK313678.1 | ||||
BC075014.2 | AAH75014.1 | |||
BC075015.2 | AAH75015.1 | |||
BX956931.1 | ||||
DQ424904.1 | ABD83669.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv7011 | Chr.X:100857971 - 100890941 on Build GRCh38 | Insertion | XKRX NOX1 HNRNPA1P27 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |