Assay Details
Target Gene Details
Entrez Gene ID: | 117154 |
Gene Name: | dachshund family transcription factor 2 |
Gene Aliases: |
- |
Location: |
Chr.X:86148451-86832602 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DACH2 | NM_001139514.1 | NP_001132986.1 | ||
NM_053281.3 | NP_444511.1 | |||
XM_011530846.2 | XP_011529148.1 | |||
XM_011530847.2 | XP_011529149.1 | |||
XM_011530848.2 | XP_011529150.1 | |||
XM_017029254.1 | XP_016884743.1 | |||
XM_017029255.1 | XP_016884744.1 | |||
XM_017029256.1 | XP_016884745.1 | |||
AF428101.1 | AAL26561.1 | |||
AK091842.1 | BAC03761.1 | |||
AK290596.1 | ||||
BC114950.1 | AAI14951.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2740276 | Chr.X:85399134 - 86284474 on Build GRCh38 | Deletion | MIR361 CHM DACH2 MIR1321 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |