Assay Details
Target Gene Details
Entrez Gene ID: | 169966 |
Gene Name: | family with sequence similarity 46 member D |
Gene Aliases: |
CT1.26, CT112 |
Location: |
Chr.X:80335504-80445311 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM46D | NM_001170574.1 | NP_001164045.1 | ||
AK128147.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |