Assay Details
Target Gene Details
Entrez Gene ID: | 266629 |
Gene Name: | SEC14 like lipid binding 3 |
Gene Aliases: |
TAP2 |
Location: |
Chr.22:30447959-30472047 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SEC14L3 | XM_011530128.2 | XP_011528430.1 | ||
AK131358.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv588886 | Chr.22:30440326 - 30472431 on Build GRCh38 | Loss | SEC14L3 |
nsv829171 | Chr.22:30202041 - 30647591 on Build GRCh38 | Loss | LIF RNF215 OSM TCN2 TBC1D10A MGC20647 SEC14L2 SDC4P SEC14L3 MTFP1 GATSL3 SLC35E4 GAL3ST1 SF3A1 CCDC157 SEC14L4 SEC14L6 HORMAD2 KIAA1656 PES1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |