Assay Details
Target Gene Details
Entrez Gene ID: | 5435 |
Gene Name: | RNA polymerase II subunit F |
Gene Aliases: |
HRBP14.4, POLRF, RPABC14.4, RPABC2, RPB14.4, RPB6, RPC15 |
Location: |
Chr.22:37953663-38041915 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POLR2F | NM_001301129.1 | NP_001288058.1 | ||
NM_001301130.1 | NP_001288059.1 | |||
NM_001301131.1 | NP_001288060.1 | |||
NM_021974.4 | NP_068809.1 | |||
NR_125371.1 | ||||
XM_017028832.1 | XP_016884321.1 | |||
AK094034.1 | ||||
AK311128.1 | ||||
AK311831.1 | ||||
BC003582.1 | AAH03582.1 | |||
BP195499.1 | ||||
CR456546.1 | CAG30432.1 | |||
CR457021.1 | CAG33302.1 | |||
DA083933.1 | ||||
DA203006.1 | ||||
DA372532.1 | ||||
DN989996.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3647723 | Chr.22:37960931 - 37962775 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)