Assay Details
Target Gene Details
Entrez Gene ID: | 100506195 |
Gene Name: | LARGE antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.22:33724986-33750817 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LARGE-AS1 | NR_038949.1 | |||
NR_038950.1 |
Target Gene Details
Entrez Gene ID: | 9215 |
Gene Name: | LARGE xylosyl- and glucuronyltransferase 1 |
Gene Aliases: |
LARGE, MDC1D, MDDGA6, MDDGB6 |
Location: |
Chr.22:33162237-33922841 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LARGE1 | NM_004737.4 | NP_004728.1 | ||
NM_133642.3 | NP_598397.1 | |||
XM_005261831.3 | XP_005261888.1 | |||
XM_005261832.3 | XP_005261889.1 | |||
XM_011530510.2 | XP_011528812.1 | |||
XM_011530514.2 | XP_011528816.1 | |||
AB011181.3 | BAA25535.3 | |||
AJ007583.1 | CAA07571.1 | |||
AK294766.1 | ||||
AK295636.1 | ||||
BC117425.1 | ||||
BC126404.1 | ||||
CR456510.1 | CAG30396.1 | |||
KJ534877.1 | ||||
KJ535033.1 | ||||
KJ535073.1 | ||||
KJ535074.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1055791 | Chr.22:33426240 - 33763549 on Build GRCh38 | Loss | LARGE1 LARGE-AS1 MIR4764 |
nsv1060684 | Chr.22:33712476 - 33744666 on Build GRCh38 | Loss | LARGE1 LARGE-AS1 |
nsv524432 | Chr.22:33723880 - 33750381 on Build GRCh38 | Loss | LARGE1 LARGE-AS1 |
nsv459878 | Chr.22:33710759 - 33830604 on Build GRCh38 | Loss | LARGE1 LARGE-AS1 |
esv2763261 | Chr.22:33435362 - 33892401 on Build GRCh38 | Loss | LARGE1 LARGE-AS1 MIR4764 |
nsv1055203 | Chr.22:33702431 - 33800698 on Build GRCh38 | Loss | LARGE1 LARGE-AS1 |
esv3568379 | Chr.22:33458829 - 33815605 on Build GRCh38 | Loss | LARGE1 LARGE-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |