Assay Details
Target Gene Details
Entrez Gene ID: | 8897 |
Gene Name: | myotubularin related protein 3 |
Gene Aliases: |
FYVE-DSP1, ZFYVE10 |
Location: |
Chr.22:29883165-30030868 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MTMR3 | NM_021090.3 | NP_066576.1 | ||
NM_153050.2 | NP_694690.1 | |||
NM_153051.2 | NP_694691.1 | |||
XM_005261803.2 | XP_005261860.1 | |||
XM_005261804.3 | XP_005261861.1 | |||
XM_005261805.2 | XP_005261862.1 | |||
XM_005261806.2 | XP_005261863.1 | |||
XM_005261808.2 | XP_005261865.1 | |||
XM_017029034.1 | XP_016884523.1 | |||
AB002369.1 | ||||
AK289581.1 | ||||
BC142713.1 | ||||
BC151217.1 | ||||
BC152455.1 | ||||
BI462128.1 | ||||
DA126318.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3647549 | Chr.22:29892602 - 29927548 on Build GRCh38 | Loss |
![]() |
esv3568352 | Chr.22:29893171 - 29957035 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)