Assay Details
Target Gene Details
Entrez Gene ID: | 157 |
Gene Name: | G protein-coupled receptor kinase 3 |
Gene Aliases: |
ADRBK2, BARK2 |
Location: |
Chr.22:25564894-25729294 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRK3 | NM_005160.3 | NP_005151.2 | ||
AK058103.1 | ||||
AK292234.1 | ||||
AK310398.1 | ||||
BC036797.1 | AAH36797.1 | |||
BX387339.2 | ||||
X69117.1 | CAA48870.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1066595 | Chr.22:25596614 - 26071416 on Build GRCh38 | Gain | MYO18B GRK3 |
nsv517205 | Chr.22:25244661 - 25629591 on Build GRCh38 | Gain+Loss | MIR6817 LOC100652901 GRK3 LOC105372966 IGLL3P LRP5L LOC105372969 CRYBB2P1 |
nsv1058343 | Chr.22:25507736 - 25642811 on Build GRCh38 | Loss | GRK3 |
esv2758829 | Chr.22:24970834 - 25650546 on Build GRCh38 | Gain+Loss | MIR6817 LOC100652901 GRK3 LOC100128531 KIAA1671 CRYBB3 CRYBB2 LOC105372966 IGLL3P LRP5L LOC105372969 CRYBB2P1 |
esv2751952 | Chr.22:25262579 - 25754679 on Build GRCh38 | Gain | MIR6817 LOC100652901 MYO18B GRK3 LOC105372966 IGLL3P LRP5L LOC105372969 CRYBB2P1 |
esv3647478 | Chr.22:25598397 - 25702971 on Build GRCh38 | Gain | GRK3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |