Assay Details
Target Gene Details
Entrez Gene ID: | 23598 |
Gene Name: | POZ/BTB and AT hook containing zinc finger 1 |
Gene Aliases: |
MAZR, PATZ, RIAZ, ZBTB19, ZNF278, ZSG, dJ400N23 |
Location: |
Chr.22:31325804-31346263 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PATZ1 | NM_014323.2 | NP_055138.2 | ||
NM_032050.1 | NP_114439.1 | |||
NM_032052.1 | NP_114441.1 | |||
AB208787.1 | BAD92024.1 | |||
AF254083.1 | AAG09032.1 | |||
AF254084.1 | AAG09033.1 | |||
AF254085.1 | AAG09034.1 | |||
AL096880.1 | CAB51404.1 | |||
AY028384.1 | AAK19024.1 | |||
CR456613.1 | CAG30499.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834176 | Chr.22:31286440 - 31468715 on Build GRCh38 | Loss | EIF4ENIF1 LINC01521 PATZ1 PIK3IP1 PIK3IP1-AS1 DRG1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |