Assay Details
Target Gene Details
Entrez Gene ID: | 3454 |
Gene Name: | interferon alpha and beta receptor subunit 1 |
Gene Aliases: |
AVP, IFN-alpha-REC, IFNAR, IFNBR, IFRC |
Location: |
Chr.21:33324443-33360361 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IFNAR1 | NM_000629.2 | NP_000620.2 | ||
XM_005260964.2 | XP_005261021.1 | |||
XM_011529552.1 | XP_011527854.1 | |||
AK222770.1 | BAD96490.1 | |||
AK222812.1 | BAD96532.1 | |||
AK298051.1 | ||||
AK312631.1 | ||||
BC002590.2 | AAH02590.1 | |||
BC021825.1 | AAH21825.1 | |||
BG548299.1 | ||||
J03171.1 | AAA52730.1 | |||
JN543953.1 | ||||
JN543954.1 | ||||
JN543955.1 | ||||
JN543956.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834087 | Chr.21:33325890 - 33469002 on Build GRCh38 | Gain |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)