Assay Details
Target Gene Details
Entrez Gene ID: | 3460 |
Gene Name: | interferon gamma receptor 2 (interferon gamma transducer 1) |
Gene Aliases: |
AF-1, IFGR2, IFNGT1, IMD28 |
Location: |
Chr.21:33402896-33437521 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IFNGR2 | NM_005534.3 | NP_005525.2 | ||
XM_005260969.2 | XP_005261026.1 | |||
AK292246.1 | ||||
AK314005.1 | ||||
BC003624.2 | AAH03624.1 | |||
CB992729.1 | ||||
U05875.1 | AAA16955.1 | |||
U05877.1 | AAA16956.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3646904 | Chr.21:33396481 - 33409590 on Build GRCh38 | Gain | IFNGR2 |
nsv834087 | Chr.21:33325890 - 33469002 on Build GRCh38 | Gain | TMEM50B IFNGR2 IFNAR1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |